Methyl-3-bromo-4-chlorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-bromo-4-chlorobenzoate

A764867-1G A764867-250MG A764867-5G A764867-10G
76832-056EA 33.95 USD
76832-056 76832-058 76832-060 77684-702
Methyl-3-bromo-4-chlorobenzoate 98%
Methyl 3-bromo-4-chlorobenzoate

Precaución: For research use only.

Formula: C₈H₆BrClO₂
MW: 249.4890 g/mol
MDL Number: MFCD09751943
CAS Number: 107947-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR