2,5-Diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester 95%

Supplier: Ambeed
Danger

A764534-100MG A764534-1G A764534-250MG
76832-050EA 52 USD
76832-050 76832-052 76832-054
2,5-Diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester 95%
2,5-Diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₂
MDL Number: MFCD11501185
CAS Number: 1086398-02-8
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR