2-Chloro-5-isobutoxyphenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2-Chloro-5-isobutoxyphenyl)boronic acid
A763922-1G
A763922-250MG
76832-040EA
75.33
USD
76832-040
76832-042
2-Chloro-5-isobutoxyphenylboronic acid 97%
2-Chloro-5-isobutoxyphenylboronic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₄BClO₃ MW: 228.4804 g/mol |
MDL Number:
MFCD16295072 CAS Number: 1256346-11-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...