Methyl 7-fluoro-1H-indole-2-carboxylate 97%

Supplier: Ambeed
Warning

A762692-100MG A762692-250MG
76832-034EA 101.85 USD
76832-034 76832-036
Methyl 7-fluoro-1H-indole-2-carboxylate 97%
Methyl 7-fluoro-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈FNO₂
MW: 193.17 g/mol
MDL Number: MFCD15209175
CAS Number: 1158331-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR