2-Aminoethan-1-ol hemi((Z)-3'-(2-(1-(3,4-dimethylphenyl)-3-methyl-5-oxo-1,5-dihydro-4H-pyrazol-4-ylidene)hydrazinyl)-2'-hydroxy-[1,1'-biphenyl]-3-carboxylate) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Eltrombopag Olamine
A759657-100MG
A759657-10MG
A759657-1MG
A759657-5MG
A759657-250MG
A759657-5G
76832-004EA
33.95
USD
76832-004
76832-006
76832-008
76832-010
77614-346
77684-697
2-Aminoethan-1-ol hemi((Z)-3'-(2-(1-(3,4-dimethylphenyl)-3-methyl-5-oxo-1,5-dihydro-4H-pyrazol-4-ylidene)hydrazinyl)-2'-hydroxy-[1,1'-biphenyl]-3-carboxylate) 98%
2-Aminoethan-1-ol hemi((Z)-3'-(2-(1-(3,4-dimethylphenyl)-3-methyl-5-oxo-1,5-dihydro-4H-pyrazol-4-ylidene)hydrazinyl)-2'-hydroxy-[1,1'-biphenyl]-3-carboxylate)
Precaución: For research use only.
Formula:
C₂₉H₃₆N₆O₆ MW: 564.63 g/mol |
MDL Number:
MFCD22380664 CAS Number: 496775-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...