1-(Phenylsulphonyl)-6-indoleboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (1-(Phenylsulphonyl)-1H-indol-6-yl)boronic acid

A756241-1G A756241-250MG A756241-5G
76831-980EA 111 USD
76831-980 76831-982 76831-984
1-(Phenylsulphonyl)-6-indoleboronic acid 95%
1-(Phenylsulphonyl)-6-indoleboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂BNO₄S
MW: 301.1254 g/mol
MDL Number: MFCD03840611
CAS Number: 480438-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR