1-[3,5-Bis(trifluoromethyl)phenyl]-3-[(1S,2S)-(+)-2-(dimethylamino)cyclohexyl]thiourea 98%

Supplier: Ambeed
Warning

Synonyms: (S,S-TUC), 1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1S,2S)-2-(dimethylamino)cyclohexyl)thiourea

A751788-100MG A751788-1G A751788-250MG A751788-25G A751788-5G
76831-958EA 12.74 USD
76831-958 76831-960 76831-962 76831-964 76831-966
1-[3,5-Bis(trifluoromethyl)phenyl]-3-[(1S,2S)-(+)-2-(dimethylamino)cyclohexyl]thiourea 98%
1-[3,5-Bis(trifluoromethyl)phenyl]-3-[(1S,2S)-(+)-2-(dimethylamino)cyclohexyl]thiourea

Precaución: for Research Use Only

Formula: C₁₇H₂₁F₆N₃S
MW: 413.4200 g/mol
MDL Number: MFCD09834840
CAS Number: 851477-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR