4-Methoxy-3-(3-methoxypropoxy)benzenemethanol 98+%

Supplier: Ambeed
Warning

A751527-1G A751527-250MG A751527-5G
76831-956EA 6 USD
76831-956 77886-701 77886-702
4-Methoxy-3-(3-methoxypropoxy)benzenemethanol 98+%
4-Methoxy-3-(3-methoxypropoxy)benzenemethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₄
MDL Number: MFCD12768300
CAS Number: 172900-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR