Tetrahydro-1H-oxazolo[3,4-a]pyrazin-3(5H)-one hydrochloride 97%

Supplier: Ambeed
Warning

A750897-1G A750897-250MG
76831-946EA 150.65 USD
76831-946 76831-948
Tetrahydro-1H-oxazolo[3,4-a]pyrazin-3(5H)-one hydrochloride 97%
Tetrahydro-1H-oxazolo[3,4-a]pyrazin-3(5H)-one hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁ClN₂O₂
MW: 178.62 g/mol
MDL Number: MFCD09878828
CAS Number: 1020349-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR