3-Nitro-1H-pyrazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A750830-100G A750830-1G A750830-250MG
76831-940EA 657.76 USD
76831-940 76831-942 76831-944
3-Nitro-1H-pyrazole-5-carboxylic acid 97%
3-Nitro-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₃N₃O₄
MW: 157.08 g/mol
MDL Number: MFCD01871089
CAS Number: 925646-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR