Diethyl 4-hydroxypyridine-2,6-dicarboxylate 95%

Supplier: Ambeed
Warning

A749568-100MG A749568-1G A749568-250MG A749568-5G
76831-932EA 12 USD
76831-932 76831-934 76831-936 76831-938
Diethyl 4-hydroxypyridine-2,6-dicarboxylate 95%
Diethyl 4-hydroxypyridine-2,6-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₅
MDL Number: MFCD13189672
CAS Number: 68631-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR