(4-Methylpyridin-2-yl)methylamine 97%

Supplier: Ambeed
Danger

Synonyms: 2-(Aminomethyl)-4-methylpyridine, (4-Methylpyridin-2-yl)methanamine

A749412-250MG A749412-100MG
76831-930EA 108.22 USD
76831-930 77614-306
(4-Methylpyridin-2-yl)methylamine 97%
(4-Methylpyridin-2-yl)methylamine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD06213857
CAS Number: 129768-95-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR