Ethyl benzo[d]oxazole-6-carboxylate 95+%

Supplier: Ambeed
Warning

A748380-100MG A748380-1G A748380-250MG
76831-924EA 15.92 USD
76831-924 76831-926 76831-928
Ethyl benzo[d]oxazole-6-carboxylate 95+%
Ethyl benzo[d]oxazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MW: 191.18 g/mol
MDL Number: MFCD16036195
CAS Number: 1355171-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR