Sodium 3-(trifluoromethyl)benzoate 96%

Supplier: Ambeed
Warning

A745969-100G A745969-25G A745969-5G
76831-910EA 169.75 USD
76831-910 76831-912 76831-914
Sodium 3-(trifluoromethyl)benzoate 96%
Sodium 3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₈H₄F₃NaO₂
MW: 212.10 g/mol
MDL Number: MFCD01679515
CAS Number: 69226-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR