1-Boc-3-methoxycarbonylmethyleneazetidine 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-(2-methoxy-2-oxoethylidene)azetidine-1-carboxylate

A744496-1G A744496-250MG
76831-892EA 47.75 USD
76831-892 76831-894
1-Boc-3-methoxycarbonylmethyleneazetidine 98%
1-Boc-3-methoxycarbonylmethyleneazetidine

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₄
MW: 227.26 g/mol
MDL Number: MFCD13190429
CAS Number: 1105662-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR