3-Bromo-4-chlorobenzylamine 97%

Supplier: Ambeed
Warning

A742685-1G A742685-250MG
76831-888EA 36 USD
76831-888 77886-688
3-Bromo-4-chlorobenzylamine 97%
3-Bromo-4-chlorobenzylamine

Precaución: for Research Use Only

Formula: C₇H₇BrClN
MDL Number: MFCD18390315
CAS Number: 849367-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR