(3-Chloro-5-(pyrrolidin-1-yl)phenyl)boronic acid 96%

Supplier: Ambeed
Warning

A742362-100MG A742362-1G A742362-250MG
76831-882EA 25 USD
76831-882 76831-884 76831-886
(3-Chloro-5-(pyrrolidin-1-yl)phenyl)boronic acid 96%
(3-Chloro-5-(pyrrolidin-1-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BClNO₂
MW: 225.48 g/mol
MDL Number: MFCD18783126
CAS Number: 1402238-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR