(S)-1,3-Dimethylpiperazine 95%

Supplier: Ambeed
Warning

A740637-100MG A740637-1G A740637-250MG A740637-25G A740637-5G
76831-872EA 45.62 USD
76831-872 76831-874 76831-876 76831-878 76831-880
(S)-1,3-Dimethylpiperazine 95%
(S)-1,3-Dimethylpiperazine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MW: 114.18 g/mol
MDL Number: MFCD11114436
CAS Number: 1152367-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR