N1-Methylbenzene-1,3-diamine 95%

Supplier: Ambeed
Warning

A739590-1G A739590-250MG A739590-5G
76831-868EA 60 USD
76831-868 76831-870 77886-684
N1-Methylbenzene-1,3-diamine 95%
N1-Methylbenzene-1,3-diamine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MDL Number: MFCD11132708
CAS Number: 50617-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR