(11bS)-2,6-Bis(3,5-dimethylphenyl)-4-hydroxy-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 98%
Supplier: Ambeed
|
Warning
|
A739100-100MG
A739100-1G
A739100-250MG
76831-862EA
103.97
USD
76831-862
76831-864
76831-866
(11bS)-2,6-Bis(3,5-dimethylphenyl)-4-hydroxy-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 98%
(11bS)-2,6-Bis(3,5-dimethylphenyl)-4-hydroxy-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide
Precaución: for Research Use Only
Formula:
C₃₆H₃₇O₄P MW: 564.65 g/mol |
CAS Number:
2757287-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...