4-Benzylcarbamoyl-3-chlorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Benzylcarbamoyl)-3-chlorophenyl)boronic acid, 3-Chloro-4-(N-benzylcarbamoyl)phenylboronic acid

A739020-1G A739020-250MG A739020-5G
76831-856EA 75.99 USD
76831-856 76831-858 76831-860
4-Benzylcarbamoyl-3-chlorophenylboronic acid 98%
4-Benzylcarbamoyl-3-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₃BClNO₃
MDL Number: MFCD07363769
CAS Number: 850589-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR