2-Bromo-4,4'-di-tert-butyl-1,1'-biphenyl 99%

Supplier: Ambeed
Warning

A738815-100G A738815-25G A738815-5G
76831-852EA 134 USD
76831-852 76831-854 77886-683
2-Bromo-4,4'-di-tert-butyl-1,1'-biphenyl 99%
2-Bromo-4,4'-di-tert-butyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₂₀H₂₅Br
MDL Number: MFCD20921696
CAS Number: 70728-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR