1-(2-Chloroethyl)-2-pyrrolidone 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2-chloroethyl)pyrrolidin-2-one, N-(beta-chloroethyl)-pyrrolidinone, 2-Pyrrolidinone, 1-(2-chloroethyl)-

A738792-250MG A738792-1G A738792-5G
76831-850EA 25 USD
76831-850 77614-276 77614-278
1-(2-Chloroethyl)-2-pyrrolidone 95%
1-(2-Chloroethyl)-2-pyrrolidone

Precaución: for Research Use Only

Formula: C₆H₁₀ClNO
MW: 147.6 g/mol
MDL Number: MFCD00169064
CAS Number: 51333-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR