3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-[3-(4-Methyl-1-piperazinylmethyl)phenyl]-1,3,2-dioxaborolane, 1-Methyl-4-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperazine, 3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid Pinacol ester, 1-Methyl-4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperazine

A738045-100MG A738045-1G A738045-250MG A738045-5G
76831-842EA 14 USD
76831-842 76831-844 76831-846 76831-848
3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester 96%
3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₂₉BN₂O₂
MDL Number: MFCD09266181
CAS Number: 883738-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR