((2R,3R,4R)-3-((4-Chlorobenzoyl)oxy)-4-fluoro-4-methyl-5-oxotetrahydrofuran-2-yl)methyl 4-chlorobenzoate 95%

Supplier: Ambeed
Warning

A738043-1G A738043-250MG
76831-838EA 264.17 USD
76831-838 76831-840
((2R,3R,4R)-3-((4-Chlorobenzoyl)oxy)-4-fluoro-4-methyl-5-oxotetrahydrofuran-2-yl)methyl 4-chlorobenzoate 95%
((2R,3R,4R)-3-((4-Chlorobenzoyl)oxy)-4-fluoro-4-methyl-5-oxotetrahydrofuran-2-yl)methyl 4-chlorobenzoate

Precaución: for Research Use Only

Formula: C₂₀H₁₅Cl₂FO₆
MW: 441.23 g/mol
MDL Number: MFCD21608624
CAS Number: 1294481-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR