1-Benzyloxy-4-bromo-2-chlorobenzene 98%

Supplier: Ambeed
Danger

A734856-1G A734856-5G
76831-806EA 38.2 USD
76831-806 76831-808
1-Benzyloxy-4-bromo-2-chlorobenzene 98%
1-Benzyloxy-4-bromo-2-chlorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrClO
MW: 297.5800 g/mol
MDL Number: MFCD09265138
CAS Number: 56872-27-6
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR