4-(1-Pyrrolidinylcarbonylamino)benzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyrrolidine-1-carboxamide

A734172-1G A734172-250MG A734172-5G
76831-800EA 123.07 USD
76831-800 76831-802 76831-804
4-(1-Pyrrolidinylcarbonylamino)benzeneboronic acid pinacol ester 98%
4-(1-Pyrrolidinylcarbonylamino)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₅BN₂O₃
MW: 316.21 g/mol
MDL Number: MFCD08689514
CAS Number: 874290-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR