4-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: N-Methoxy-N-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide, N-Methoxy-N-methyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

A732647-1G A732647-250MG A732647-25G A732647-5G
76831-784EA 47 USD
76831-784 76831-786 76831-788 76831-790
4-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester 98%
4-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₂BNO₄
MW: 291.16 g/mol
MDL Number: MFCD09953501
CAS Number: 1073353-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR