4-Phenylcyclohexanol 98%

Supplier: Ambeed
Warning

A732290-100MG A732290-1G A732290-250MG
76831-778EA 50.93 USD
76831-778 76831-780 76831-782
4-Phenylcyclohexanol 98%
4-Phenylcyclohexanol

Precaución: for Research Use Only

Formula: C₁₂H₁₆O
MW: 176.26 g/mol
MDL Number: MFCD00013757
CAS Number: 5437-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR