1-Benzylpiperidin-3-one hydrochloride hydrate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Benzyl-3-piperidone hydrochloride hydrate
A730344-100G
A730344-10G
A730344-1G
A730344-25G
A730344-5G
76831-766EA
262.05
USD
76831-766
76831-768
76831-770
76831-772
76831-774
1-Benzylpiperidin-3-one hydrochloride hydrate 97%
1-Benzylpiperidin-3-one hydrochloride hydrate
Precaución: for Research Use Only
Formula:
C₁₂H₁₅NO.HCl.xH₂O MW: 225.72 g/mol (anhy) |
MDL Number:
MFCD00150096 CAS Number: 346694-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...