2′-(Dicyclohexylphosphino)acetophenone ethylene ketal 98%

Supplier: Ambeed
Warning

Synonyms: Dicyclohexyl(2-(2-methyl-1,3-dioxolan-2-yl)phenyl)phosphine

A727284-1G A727284-250MG
76831-760EA 121 USD
76831-760 77886-660
2′-(Dicyclohexylphosphino)acetophenone ethylene ketal 98%
2′-(Dicyclohexylphosphino)acetophenone ethylene ketal

Precaución: for Research Use Only

Formula: C₂₂H₃₃O₂P
MW: 360.4700 g/mol
MDL Number: MFCD08705257
CAS Number: 221187-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR