5-Bromo-6-chloro-2-(methylthio)pyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A725909-1G A725909-250MG
76831-756EA 603 USD
76831-756 76831-758
5-Bromo-6-chloro-2-(methylthio)pyrimidin-4-amine 95%
5-Bromo-6-chloro-2-(methylthio)pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₅H₅BrClN₃S
MW: 254.53 g/mol
MDL Number: MFCD16988271
CAS Number: 63931-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR