trans-2-(3-Fluorophenyl)vinylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (E)-(3-Fluorostyryl)boronic acid

A725174-1G A725174-250MG A725174-5G
76831-746EA 148.53 USD
76831-746 76831-748 76831-750
trans-2-(3-Fluorophenyl)vinylboronic acid 95%
trans-2-(3-Fluorophenyl)vinylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BFO₂
MW: 165.957 g/mol
MDL Number: MFCD06008297
CAS Number: 849062-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR