5-Iodo-3-(trifluoromethyl)pyridin-2-amine 97%

Supplier: Ambeed
Warning

A717649-100MG A717649-1G
76831-742EA 7 USD
76831-742 76831-744
5-Iodo-3-(trifluoromethyl)pyridin-2-amine 97%
5-Iodo-3-(trifluoromethyl)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₄F₃IN₂
MDL Number: MFCD16250855
CAS Number: 911112-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR