2,6-Dichloro-1,5-naphthyridine 95%

Supplier: Ambeed
Danger

A715699-1G A715699-250MG
76831-732EA 264.17 USD
76831-732 77614-200
2,6-Dichloro-1,5-naphthyridine 95%
2,6-Dichloro-1,5-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₄Cl₂N₂
MW: 199.03 g/mol
MDL Number: MFCD03931483
CAS Number: 27017-66-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR