1-(tert-Butyl) 2-methyl (S)-aziridine-1,2-dicarboxylate 95%

Supplier: Ambeed
Warning

A715306-100MG A715306-1G A715306-250MG A715306-5G
76831-726EA 47.75 USD
76831-726 76831-728 76831-730 77552-190
1-(tert-Butyl) 2-methyl (S)-aziridine-1,2-dicarboxylate 95%
1-(tert-Butyl) 2-methyl (S)-aziridine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₅NO₄
MW: 201.22 g/mol
MDL Number: MFCD10566310
CAS Number: 126496-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR