Ethyl 3-(3,5-difluorophenyl)-3-oxopropanoate 95%

Supplier: Ambeed
Warning

A713098-100MG A713098-250MG
76831-716EA 45.62 USD
76831-716 76831-718
Ethyl 3-(3,5-difluorophenyl)-3-oxopropanoate 95%
Ethyl 3-(3,5-difluorophenyl)-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₀F₂O₃
MW: 228.19 g/mol
MDL Number: MFCD07783525
CAS Number: 359424-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR