Methyl 2-(4-amino-3-nitrophenyl)acetate 98%

Supplier: Ambeed
Warning

A712970-100MG A712970-1G A712970-250MG A712970-5G
76831-708EA 22.28 USD
76831-708 76831-710 76831-712 76831-714
Methyl 2-(4-amino-3-nitrophenyl)acetate 98%
Methyl 2-(4-amino-3-nitrophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₄
MW: 210.18 g/mol
MDL Number: MFCD16620323
CAS Number: 28694-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR