(2-Methyl-5-nitrophenyl)methanol 97%

Supplier: Ambeed
Warning

A710698-100MG A710698-250MG
76831-700EA 10.61 USD
76831-700 76831-702
(2-Methyl-5-nitrophenyl)methanol 97%
(2-Methyl-5-nitrophenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD00229932
CAS Number: 22474-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR