(4-Bromo-2,5-dimethoxyphenyl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-2,5-dimethoxyphenylboronic acid
A709088-1G
A709088-250MG
A709088-5G
76831-696EA
57
USD
76831-696
76831-698
77886-647
(4-Bromo-2,5-dimethoxyphenyl)boronic acid 98%
(4-Bromo-2,5-dimethoxyphenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₈H₁₀BBrO₄ |
MDL Number:
MFCD07778019 CAS Number: 950846-26-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...