4-Oxo-4-((3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)amino)butanoic acid 96%

Supplier: Ambeed
Warning

A708301-1G A708301-250MG A708301-5G
76831-678EA 54.11 USD
76831-678 76831-680 76831-682
4-Oxo-4-((3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)amino)butanoic acid 96%
4-Oxo-4-((3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)amino)butanoic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₂BNO₅
MW: 319.16 g/mol
MDL Number: MFCD06795662
CAS Number: 1030269-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR