JK 184 95%

Supplier: Ambeed
Warning

Synonyms: JK184, N-(4-Ethoxyphenyl)-4-(2-methylimidazo[1,2-a]pyridin-3-yl)thiazol-2-amine

A706302-100MG A706302-10MG A706302-1MG A706302-25MG A706302-50MG A706302-5MG
76831-666EA 137.92 USD
76831-666 76831-668 76831-670 76831-672 76831-674 76831-676
JK 184 95%
JK 184

Precaución: for Research Use Only

Formula: C₁₉H₁₈N₄OS
MW: 350.4374 g/mol
MDL Number: MFCD01044465
CAS Number: 315703-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR