2-(Trifluoromethyl)phenylboronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,4,5,5-Tetramethyl-2-(2-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane
A705386-1G
A705386-250MG
A705386-25G
A705386-5G
76831-658EA
27.59
USD
76831-658
76831-660
76831-662
76831-664
2-(Trifluoromethyl)phenylboronic acid pinacol ester 98%
2-(Trifluoromethyl)phenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₁₆BF₃O₂ MW: 272.0712 g/mol |
MDL Number:
MFCD06795676 CAS Number: 1073339-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...