Methyl 1,4-dihydroxy-2-naphthoate 99%, Ambeed.Inc

Supplier: Ambeed
Warning

A704843-1G A704843-25G A704843-5G
76831-654EA 16.98 USD
76831-654 76831-656 77684-617
Methyl 1,4-dihydroxy-2-naphthoate 99%, Ambeed.Inc
Methyl 1,4-dihydroxy-2-naphthoate

Precaución: For research use only.

Formula: C₁₂H₁₀O₄
MW: 218.21 g/mol
MDL Number: MFCD00573066
CAS Number: 77060-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR