2-Benzyloxy-5-pyrimidineboronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 2-(Benzyloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

A703463-100MG A703463-1G A703463-250MG
76831-646EA 37.14 USD
76831-646 76831-648 76831-650
2-Benzyloxy-5-pyrimidineboronic acid pinacol ester 96%
2-Benzyloxy-5-pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₁BN₂O₃
MW: 312.1700 g/mol
MDL Number: MFCD11878343
CAS Number: 1218791-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR