Ethyl 5-aminoisoxazole-3-carboxylate 95%

Supplier: Ambeed
Warning

A700579-250MG A700579-1G A700579-100MG
76831-632EA 56 USD
76831-632 77573-262 77614-172
Ethyl 5-aminoisoxazole-3-carboxylate 95%
Ethyl 5-aminoisoxazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₃
MDL Number: MFCD13174972
CAS Number: 485807-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR