4-Methylpiperidin-4-amine dihydrochloride 95%

Supplier: Ambeed
Warning

A699489-100MG A699489-1G
76831-626EA 117 USD
76831-626 76831-628
4-Methylpiperidin-4-amine dihydrochloride 95%
4-Methylpiperidin-4-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆Cl₂N₂
MW: 187.11 g/mol
MDL Number: MFCD15474937
CAS Number: 483366-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR