2-(Cyclopropylamino)-5-pyrimidineboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: N-Cyclopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

A698868-100MG A698868-1G A698868-250MG
76831-614EA 36.08 USD
76831-614 76831-616 76831-618
2-(Cyclopropylamino)-5-pyrimidineboronic acid pinacol ester 95%
2-(Cyclopropylamino)-5-pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₀BN₃O₂
MW: 261.1300 g/mol
MDL Number: MFCD07375178
CAS Number: 1218789-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR