Itaconic acid 99% + stabilized
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Methylenesuccinic acid, Methylenesuccinic acid
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC)
A697659-1000G
A697659-100G
A697659-25G
A697659-500G
76831-606EA
26.53
USD
76831-606
76831-608
76831-610
76831-612
Itaconic acid 99% + stabilized
Itaconic acid
Precaución: for Research Use Only
Formula:
C₅H₆O₄ MW: 130.1 g/mol |
MDL Number:
MFCD00004260 CAS Number: 97-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...