3-Bromo-1-tosyl-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: Ambeed
Warning

A694754-10G A694754-1G A694754-5G
76831-588EA 186.72 USD
76831-588 76831-590 76831-592
3-Bromo-1-tosyl-1H-pyrrolo[2,3-b]pyridine 97%
3-Bromo-1-tosyl-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrN₂O₂S
MW: 351.21 g/mol
MDL Number: MFCD10697518
CAS Number: 226085-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR